Order Entry
Austria
ContactUsLinkComponent
SYBR® Green I staining reagent DNA-free
SYBR® Green I staining reagent DNA-free
Catalog # A8511.50625
Supplier:  PanReac AppliChem
CAS Number:  
undefined
SYBR® Green I staining reagent DNA-free
Catalog # A8511.50625
Supplier:  PanReac AppliChem
CAS Number:  

Specifications

  • Pk:
    5
  • Seq:
    0005
  • Environmentally Preferable:
  • Pk Info:
    5 x 0,625 ml

Specifications

Specification Test Results

  • Bacterial DNA:
    not detectable (min. 40 PCR cycles)

Specification Test Results

About this item

Synonyms: Thiazole Green
 :
 :509.73 g/mol
Storage Temperature:Freezer
CAS Number:163795-75-3
Formula:C₃₂H₃₇N₄S
MDL Number:MFCD00284715