Microplates, 96-well, no bottom
Supplier: Greiner Bio-One
Certificates
About this item
These 96-well plates are made from virgin polystyrene resin and moulded with no bottom.
- Chimney-well design to help prevent cross-contamination
Specifications
- Catalog No:
- GREI655000-06_40
- GREI655000_40
- Environmentally Preferable:
- Umweltschonende Herstellung
- Umweltschonende Herstellung
- Umweltzertifizierung:
- ISO 14001 (241002); ISO 50001 (241002)
- ISO 14001 (241002); ISO 50001 (241002)