Order Entry
Austria
Orders LinkContactUsLinkComponent
ORP test solution

ORP test solution

ORP test solution, 200 mV, Plastic bottle, 500 mL
Catalog # HACH25M2A1001-115
Supplier:   Hach
ORP test solution
Catalog # HACH25M2A1001-115
Supplier:  Hach
CAS Number:  
ORP test solution
ORP test solution

ORP test solution, 200 mV, Plastic bottle, 500 mL

CAS Number:  
Price
76,30 €
Log in for availability
1 * 500 mL
Product Details & Documents

ORP (Oxidation Reduction Potential) reference solution used for ORP sensor calibration.

Documents

Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    500 mL
  • Description:
    ORP test solution, 200 mV

Specifications