Push button lockout devices
Supplier: Brady
Jump to:
About this item
These lockout devices prevent the unauthorised use of push button emergency devices.
- With standard cover
- Made from clear PC, with translucent red cover
- Suitable for 16, 22 or 30 mm push buttons
11 Options Available Below
Product Details & Documents
These lockout devices prevent the unauthorised use of push button emergency devices.
- With standard cover
- Made from clear PC, with translucent red cover
- Suitable for 16, 22 or 30 mm push buttons
Product Family Options
Product Information
- TypeColourAvailabilityPrice
Bases