Orders LinkContactUsLinkComponent
Draining racks
Draining racks
With 24 rods (95 mm) and 20 arches (2×110 mm and 3×70 mm), 420×170×610 mm
Catalog # 149-1351
Draining racks
Catalog # 149-1351
Draining racks
With 24 rods (95 mm) and 20 arches (2×110 mm and 3×70 mm), 420×170×610 mm
Price
334,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Wire, PE-coated.
  • Very good chemical resistance
  • Very good temperature resistance
  • Easy to clean
It is designed with free-standing or for wall-mounting and with drip pan. The feet are protected by plastic caps.
Specifications
  • Description:
    With 24 rods (95 mm) and 20 arches (2×110 mm and 3×70 mm)
  • W×D×H:
    420×170×610 mm