Order Entry
Austria
ContactUsLinkComponent
Organic reference standards, ginsenoside Rb1, Ph. Eur.
Organic reference standards, ginsenoside Rb1, Ph. Eur.
Catalog # EDQMY0001347
Supplier:  EDQM
CAS Number:  
Organic reference standards, ginsenoside Rb1, Ph. Eur.
Catalog # EDQMY0001347
Supplier:  EDQM
CAS Number:  

Specifications

  • Pk:
    30 mg
  • Standards:
    Reference standard
  • CAS No.:
    41753-43-9
  • Description:
    Ginsenoside Rb1
  • Formula:
    C₅₄H₉₂O₂₃
  • Description:
    Ginsenoside Rb1

Specifications

About this item