Crucibles, Tammann, aluminum oxide
Supplier: Thermo Fisher Scientific
Certificates
About this item
These crucibles according to Tammann are manufactured from aluminium oxide. They display superior characteristics both at elevated temperature and at high frequencies, and are resistant to corrosive liquids and gases. Oxide ceramics are also inert to oxidation and are not subject to radiation damage.
Frequently Bought Together