Orders LinkContactUsLinkComponent
Microscope slides for printers
Pink
  KLINKP-SIL-3059
 :   KLINIPATH
Microscope slides for printers
  KLINKP-SIL-3059
 :  KLINIPATH
Microscope slides for printers
Pink
270,00 €
1 * 2.500 ST
Microscope slides for slide printers.
  • Frosted ends
  • 90° ground edges
  • Finished clipped corners of 1 mm
Allowing for easy handling of automated slide printers and digital slide imagers in slide racks.
  • Colour:
    Pink
  • Seq:
    11