ContactUsLinkComponent
Vydac 201TP C18, HPLC columns
 :  Avantor
undefined
Vydac 201TP C18, HPLC columns
 :  Avantor
  • Catalog No:
  • 201TP52
  • 554-7583
  • 201TP5410
  • 201TP104
  • HICH201GK52/N
  • 201TP5415
  • 201TP5215
  • 201TP5315
  • HICH201GK104/N
  • HICH201GD104/N
  • 201TP5203
  • 201GD52/N
  • 201GD54/N
  • 201GK54/N
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Carbon load:
  • 8%
  • 8%
  • 8%
  • 8%
  • 8%
  • 8%
  • 8%
  • 8%
  • Endcapped:
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • USP code:
  • L1
  • L1
  • L1
  • L1
  • L1
  • L1
  • L1
  • L1
  • Environmental certification:
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
Guard holders and accessories
Frequently Bought Together