Order Entry
Austria
ContactUsLinkComponent
Vydac 202TP C18-HL, HPLC columns
Vydac 202TP C18-HL, HPLC columns
Catalog # 554-7582
Supplier:  Avantor
   Low Manufacturing Impact
undefined
Vydac 202TP C18-HL, HPLC columns
Catalog # 554-7582
Supplier:  Avantor
   Low Manufacturing Impact

Specifications

  • Column Format:
    Analytical
  • Ø int.:
    4,60 mm
  • Particle Size:
    5 µm
  • Pore size:
    300 Å
  • Length:
    250 mm
  • Seq:
    02
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    ISO 14001:2015 (530840 UM15)
  • Endcapped:
    No
  • USP code:
    L1
  • Surface area:
    60 - 90 m²/g
  • Carbon load:
    9%
  • Column hardware:
    Stainless steel
  • Phase:
    C18-HL
  • Packing material:
    Silica
  • Pk:
    1

Specifications

About this item

Vydac TP is the first range of wide pore media phases developed in the Vydac range and it is the industry's standard for peptide and protein separations. The large pores of the 300 Å TP silica give polypeptide molecules complete access to the interior of the silica pores.

  • Long column lifetime and negligible phase leaching
  • Reliable protein purifications, scalable from analytical to preparative scale
  • Referenced in a large number of patents and publications
  • USP L1

Higher carbon load than 201TP. Developed for the separation of PAHs.

* Use with holder 80101/N and column coupler HI-081.

Vydac is a registered trademark of W.R.Grace and Co.