Handheld digital microscopes, Dino-Lite Special Light, UV
Supplier: Dino-Lite / IDCP B.V.
Certificates
About this item
These handheld microscopes feature ultraviolet (UV) light or a combination between UV and white light.
- High speed USB 2.0 output
- Metal housing or composite housing; metal models are ESD-safe
- Edge series models with high-quality optics provide a very sharp, bright and natural colour image with very low aberration and vignetting
- Wireless streaming to a tablet, smartphone, or computer is possible in combination with WF-10 unit (to be ordered separately)
- Integrated polariser greatly reduces glare on reflective surfaces (some models only)
- Exchangeable caps for more flexibility (some models only)
Delivery: Supplied with carry pouch, software CD (DinoCapture 2.0 (Windows), DinoXcope (Mac OS)), user manual, Quick guide and calibration target.
Frequently Bought Together