Order Entry
Austria
ContactUsLinkComponent
Anti-IgG Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anti-IgG Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # 35-8077-M002
Supplier:  Tonbo Biosciences
Anti-IgG Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # 35-8077-M002
Supplier:  Tonbo Biosciences

Specifications

  • Antibody type:
    Secondary
  • Antigen name:
    Immunoglobulin G
  • Antigen symbol:
    IgG
  • Clonality:
    Polyclonal
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Reactivity:
    Mouse
  • Epitope:
    Heavy and light chain
  • Cross adsorption:
    Yes
  • Storage temperature:
    Store at 2-8 °C
  • Pk:
    2 mg

Specifications

About this item

Rabbit anti-Mouse IgG (H&L) - Affinity Pure, FITC Conjugate, min x w/human serum proteins

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope: Heavy and light chain
Host: Rabbit
Isotype:
Reactivity: Mouse