Order Entry
Austria
ContactUsLinkComponent
 
Anti-IgM Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
  35-8065-M002
Anti-IgM Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
  35-8065-M002

 

  • Antibody type:
    Secondary
  • Antigen name:
    Immunoglobulin M mu
  • Antigen symbol:
    IgM
  • Clonality:
    Polyclonal
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Goat
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Reactivity:
    Mouse
  • Epitope:
    µ chain
  • Storage temperature:
    Store at 2-8 °C
  • Pk:
    2 mg

 

 

Goat anti-Mouse IgM (mu chain) - Affinity Pure, FITC Conjugate

Type: Secondary
Antigen: IgM
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope: µ chain
Host: Goat
Isotype:
Reactivity: Mouse