Order Entry
Austria
ContactUsLinkComponent
Anti-IgG Goat Polyclonal Antibody
Anti-IgG Goat Polyclonal Antibody
Catalog # 70-8042-M002
Supplier:  Tonbo Biosciences
Anti-IgG Goat Polyclonal Antibody
Catalog # 70-8042-M002
Supplier:  Tonbo Biosciences

Specifications

  • Antibody type:
    Secondary
  • Antigen name:
    Immunoglobulin G
  • Antigen symbol:
    IgG
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Goat
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Mouse
  • Western blot:
    Yes
  • Epitope:
    Heavy and light chain
  • Storage temperature:
    Store at 2-8 °C
  • Pk:
    2 mg

Specifications

About this item

Goat anti-Mouse IgG (H&L) - Affinity Pure

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Heavy and light chain
Host: Goat
Isotype:
Reactivity: Mouse