Order Entry
Austria
ContactUsLinkComponent
Anti-IgM Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anti-IgM Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # 35-8030-M001
Supplier:  Tonbo Biosciences
Anti-IgM Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # 35-8030-M001
Supplier:  Tonbo Biosciences

Specifications

  • Antibody type:
    Secondary
  • Antigen name:
    Immunoglobulin M mu
  • Antigen symbol:
    IgM
  • Clonality:
    Polyclonal
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Goat
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Reactivity:
    Human
  • Epitope:
    µ chain
  • Cross adsorption:
    Yes
  • Storage temperature:
    Store at 2-8 °C
  • Pk:
    1 mg

Specifications

About this item

Goat anti-Human IgM (mu chain) - Affinity Pure, FITC Conjugate, min x w/human IgA + IgG

Type: Secondary
Antigen: IgM
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope: µ chain
Host: Goat
Isotype:
Reactivity: Human