Shaking water baths, Precision™
Supplier: THERMO ELECTRON LED
Certificates
About this item
Precision™ water baths support a range of sensitive life science and QA/QC applications, from warming fragile reagents to tissue culturing and genetics sequencing.
- Coil-free interior for easy cleaning
- Four temperature and shaking speed presets
- Audible alarms protect work
- Auto-on and auto-off detectors
- Low-fluid detection
- Monitor with icon based graphic display
The shallow form shaking bath has a removable tray that allows it to be used with smaller sample containers. Dubnoff shaking water baths are designed specifically for applications that require samples to be incubated in a controlled atmosphere.
Delivery information: Supplied with one large and two small gassing hoods, stainless steel gable cover and shaking tray. Please note flask clips and fasteners must be ordered separately.
Specifications
- Catalog No:
- HAKATSSWB15
- HAKATSSWB15S
- HAKATSSWB27
- HAKATSDUB15
- Model:
- Precision™ SWB 15
- Precision™ SWB 15S
- Precision™ SWB 27
- Precision™ DUB 15
- Temperature range:
- 5...100 °C
- 5...100 °C
- 5...100 °C
- 5...100 °C
- Temperature stability:
- ±0,1 %
- ±0,1 %
- ±0,1 %
- ±0,1 %
- Temperature uniformity:
- ±0,5 °C
- ±0,5 °C
- ±0,5 °C
- ±0,5 °C
- W×D×H:
- 632×394×249 mm
- 632×394×249 mm
- 938×394×249 mm
- 636×396×250 mm
Specifications
Frequently Bought Together