Orders LinkContactUsLinkComponent
Pipette holder, SINGLE™
Pipette holder, SINGLE™
Pipette holder, SINGLE™, for one Gilson pipette, can be stucked to any smooth, dry surface or fastened with screws, White
Catalog # GILSF161406
Supplier:   GILSON
Pipette holder, SINGLE™
Catalog # GILSF161406
Supplier:  GILSON
Pipette holder, SINGLE™
Pipette holder, SINGLE™
Pipette holder, SINGLE™, for one Gilson pipette, can be stucked to any smooth, dry surface or fastened with screws, White
Price
19,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
This convenient pipette holder has been designed to hold one single or multi channel pipette.
  • Made of PC: Resistant to aggressive chemicals and UV light
  • Adheres strongly to any smooth, dry surface or may be fastened with screws
  • Protects valuable instruments from shocks and contamination
  • Pipettes hang vertically to prevent liquids from running inside the shaft
  • Space saving: Adapted to limited work bench space
Specifications
  • Colour:
    White
  • Description:
    Pipette holder, SINGLE™, for one Gilson pipette, can be stucked to any smooth, dry surface or fastened with screws