ContactUsLinkComponent
Pipette stand for Gilson pipettes, TRIO™
Pipette stand for Gilson pipettes, TRIO™
Catalog # GILSF161405
Supplier:  GILSON
undefined
Pipette stand for Gilson pipettes, TRIO™
Catalog # GILSF161405
Supplier:  GILSON
Specifications
  • Colour:
    Translucent
  • Description:
    Angled pipette stand, TRIO™, for 3 Gilson pipettes (except Pipetman Concept®)
  • Environmentally Preferable:
  • Pk:
    1
Specifications
About this item
This handy stand with its angled, transparent design holds up to three single or multi channel Gilson pipettes.
  • Made of Temperlite material: Very shock resistant, can be used in chemically aggressive environments
  • Protects valuable instruments from shocks and contamination
  • Pipettes hang vertically to prevent liquids from running inside the shaft
  • Maintain visibility: Its transparent body allows users to see the objects placed behind
  • Space saving for a neat, tidy and organized work bench