ContactUsLinkComponent
Pipette stand for Gilson pipettes, rotary, CARROUSEL™
Pipette stand for Gilson pipettes, rotary, CARROUSEL™
Catalog # 612-1816
Supplier:  GILSON
undefined
Pipette stand for Gilson pipettes, rotary, CARROUSEL™
Catalog # 612-1816
Supplier:  GILSON
Specifications
  • Colour:
    White
  • Description:
    Rotating pipette stand, CARROUSEL™, for 7 Gilson pipettes (except Pipetman Concept®)
  • Pk:
    1
Specifications
About this item
This carousel stand holds up to seven single or multi channel Gilson pipettes.
  • Robust and stable due to its heavy base
  • Made of PP and PC: Strong resistance to aggressive chemicals
  • Protects valuable instruments from shocks and contamination
  • Pipettes hang vertically to prevent liquids from running inside the shaft
  • Rotating head gives users an easy access to the pipette of their choice
  • Space saving for a a neat, tidy and organised work bench