Pipette stand, rotary
Catalog # 613-5492
Supplier: Sartorius
Specifications
- For:6 mechanical pipettes
- Colour:White
- Description:Carousel stand for 6 mechanical pipettes
- W×D×H:290×250×60 mm
- Environmentally Preferable:Low Manufacturing Impact
- Environmental certification:Renewable energy certificate; ISO 14001
- Pk:1
Specifications
About this item
Carousel stand for six mechanical pipettes.
When the pipette is not in use, it should be stored in an upright position in order to avoid contamination from desks and benches. This compact carousel stand is ideal for saving bench space and is compatible with all mechanical pipettes.