Quick-dry foam squeegee
About this item
These twin-blade neoprene rubber squeegees are specifically designed for use on uneven and irregular surfaces. Features a durable polymer channel and flexible blades that quickly wipe surfaces dry. Lightweight, non-marring, and easy to clean with warm water.
- Light weight and non-marring
- Easy to clean with warm water
- Easy blade change system
Resistant to water, oil, grease, detergents, sanitisers, and solvents.
Note: Not recommended for abrasive surfaces.
: Polymer adjustable or fixed handle must be ordered separately.
2 Options Available Below
Product Details & Documents
These twin-blade neoprene rubber squeegees are specifically designed for use on uneven and irregular surfaces. Features a durable polymer channel and flexible blades that quickly wipe surfaces dry. Lightweight, non-marring, and easy to clean with warm water.
- Light weight and non-marring
- Easy to clean with warm water
- Easy blade change system
Resistant to water, oil, grease, detergents, sanitisers, and solvents.
Note: Not recommended for abrasive surfaces.
: Polymer adjustable or fixed handle must be ordered separately.
Product Family Options
Product Information
Handles
Product Information
- Catalog #Handle materialHandle lengthDescriptionHandle colourAvailabilityPrice
- Polymer86 - 157,5 cmAdjustable handle with long connectorWhiteLog in for availability1 * 1 ST74,60 €