ContactUsLinkComponent
Scalpel handle, Cygnetic®
Scalpel handle, Cygnetic®
Catalog # SWAN5398
Supplier:  SWANN MORTON
undefined
Scalpel handle, Cygnetic®
Catalog # SWAN5398
Supplier:  SWANN MORTON
Specifications
  • Description:
    Handle, stainless steel, non sterile, individually sleeved
  • Length:
    124 mm
  • Pk:
    1
Specifications
About this item
This stainless steel handle is designed to assist the OR nurse with safe and easy blade attachment and removal before, during and after the procedure.
  • Safe and easy blade attachment and removal
  • Simple disassembly for cleaning
  • Great strength and stability
  • Handle is non sterile, blades are sterile
Ideal for use within rigorous Orthopaedic and general surgical procedures.
Packaging: Handle is supplied individually sleeved. Blades (to be ordered separately) are supplied individually V.P.I. protected in metal foil packets.
Blades
Product Information