Order Entry
Austria
ContactUsLinkComponent
 
Calcium nitrate tetrahydrate 99.0-103.0% ACS for analysis
 
Calcium nitrate tetrahydrate 99.0-103.0% ACS for analysis

 

  • Solubility in water:
    2,660 g/l at 20 °C
  • Thermal decomposition:
    >130 °C
  • pH of 5% solution :
    5,0 - 7,0
  • Assay (Compl.):
    99,0 - 103,0 %
  • Acidity(as HNO₃):
    0,05%
  • Insoluble matter in H₂O:
    0,005%
  • Chloride (Cl) :
    0,005%
  • Ammonium (NH₄):
    0,005%
  • Sulfate (SO₄):
    0,002%
  • Nitrite (NO₂) :
    0,001%
  • Heavy Metals (ICP-OES):
    0,0005%

 

Frequently Bought Together