Order Entry
Austria
ContactUsLinkComponent
Benedict's reagent for qualitative detection of sugar in urine
Benedict's reagent for qualitative detection of sugar in urine
Catalog # 230045R
Supplier:  VWR Chemicals
CAS Number:  
undefined
Benedict's reagent for qualitative detection of sugar in urine
Catalog # 230045R
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • S.G. @ 20°C:
    1.192 - 1.195
  • Dextrose Test:
    Positive Reaction
  • Thiocyanate Test:
    Negative Reaction

Specification Test Results

About this item

 :
CAS Number:63126-89-6
MDL Number:MFCD00146068