Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Redox/ORP Standards
Oxidation-reduction (ORP) standards solutions, ZoBell's
Oxidation-reduction (ORP) standards solutions, ZoBell's
Oxidation-reduction (ORP) standards solutions, ZoBell's
Catalog # HACH2316949
Supplier:
Hach
CAS Number:
Certificates
Expand image
Oxidation-reduction (ORP) standards solutions, ZoBell's
Catalog # HACH2316949
Supplier:
Hach
CAS Number:
Certificates
Specifications
Pk:
500 mL
Description:
ZoBell's standard solution
Specifications
About this item
Product Details
Documentation
Product Details
Documentation