Test kits, photometric, TOC, LCK cuvette tests
Supplier: Hach
Certificates
About this item
Pre-dosed reagents for maximum safety.
- Precise and reliable measurement values
- Maximum safety for users, thanks to the closed system and low amounts of reagents
- Convenient, error-free dosing of the reagents
- Barcode label for automatic recognition in the photometer
- Differentiation between tests and measuring ranges by means of colour coding
Storage conditions: 15 - 25 °C
Certifications: DIN 38409-H3
Related Information
Certificates
Search by Lot NumberHach LCK Cuvette Tests
Product Literature
HACHLCK385_TDS_EN
Datasheet
Frequently Bought Together