Order Entry
Austria
ContactUsLinkComponent
Test kits, photometric, Cyanide, easily liberatable, LCK cuvette tests
Test kits, photometric, Cyanide, easily liberatable, LCK cuvette tests
Catalog # 705-3029
Supplier:  Hach
CAS Number:  
Test kits, photometric, Cyanide, easily liberatable, LCK cuvette tests
Catalog # 705-3029
Supplier:  Hach
CAS Number:  

Specifications

  • Chemical method:
    Hach Method
  • Pk:
    24 Tests
  • Description:
    Cyanide (easily liberated)
  • Seq:
    230
  • Type:
    LCK319
  • For unit:
    DR3900, DR6000, DR1900, DR2800, DR3800, DR5000
  • Range:
    0,03 - 0,35 mg/l CN

Specifications

About this item

Pre-dosed reagents for maximum safety.

  • Precise and reliable measurement values
  • Maximum safety for users, thanks to the closed system and low amounts of reagents
  • Convenient and error-free dosing of the reagents
  • Barcode label for automatic recognition in the photometer
  • Differentiation between tests and measuring ranges by means of colour-coding

Storage conditions: 2 to 8 °C (keep refrigerated)