Surgical blade, No. 3, stitch cutter
Catalog # SWAN0326
Supplier: SWANN MORTON
Specifications
- Type:No. 3
- Description:Stitch cutter blade, stainless steel, sterile
- Environmentally Preferable:
- Pk:100
Specifications
About this item
The stainless steel No. 3 or 'baby' stitch cutter is used in conjunction with the No. 3 surgical handle.
- Convenient and cost effective
- Sterile
Enables a more aseptic techinique by operating further away from the actual wound which also allows better visibility.
Fits handles 3 Graduated, 3L, 5B, 7, 9, B3 and B3L.