


Sulphuric acid 95.0-98.0% (by acidimetry) NF, ACS, J.T.Baker®
Catalog # 9661-03
Supplier: Avantor
CAS Number:
Specifications
- Pack type:Glass bottle, clear
- Pk:2,5 L
- Seq:5096
Specifications
Specification Test Results
- GMP Manufactured Product:—
- Meets N.F. Requirements:—
- Meets ACS Reagent Chemical Requirements:—
- CAUTION: For Manufacturing, processing or repackaging:—
- Bulk Pharmaceutical Chemical:—
- NF - Assay (H₂SO₄) (by acidimetry):95.0 - 98.0 %
- NF - Arsenic (As):≤1 ppm
- NF - Chloride (Cl):≤0.005 %
- NF - Identification:Passes Test
- NF - Reducing Substances:Passes Test
- NF - Residue on Ignition:≤0.005 %
- Ammonium (NH₄):≤1 ppm
- Appearance:Passes Test
- Assay (H₂SO₄):95.0 - 98.0 %
- Chloride (Cl):≤0.1 ppm
- Color (APHA):≤10
- Heavy Metals (as Pb):≤500.0 ppb
- Nitrate (NO₃):≤0.5 ppm
- Residue after Ignition:≤4 ppm
- Specific Gravity at 60°/60°F:≥1.84
- Substances Reducing Permanganate (as SO₂):≤2 ppm
- Trace Impurities - Aluminum (Al):≤200.0 ppb
- Trace Impurities - Arsenic (As):≤0.004 ppm
- Trace Impurities - Calcium (Ca):≤200.0 ppb
- Trace Impurities - Chromium (Cr):≤100.0 ppb
- Trace Impurities - Copper (Cu):≤100.0 ppb
- Trace Impurities - Gold (Au):≤100.0 ppb
- Trace Impurities - Iron (Fe):≤200.0 ppb
- Trace Impurities - Lead (Pb):≤200.0 ppb
- Trace Impurities - Magnesium (Mg):≤200.0 ppb
- Trace Impurities - Manganese (Mn):≤100.0 ppb
- Trace Impurities - Mercury (Hg):≤5.0 ppb
- Trace Impurities - Nickel (Ni):≤200.0 ppb
- Trace Impurities - Potassium (K):≤300.0 ppb
- Trace Impurities - Sodium (Na):≤300 ppb
- Trace Impurities - Tin (Sn):≤200.0 ppb
- Trace Impurities - Titanium (Ti):≤200.0 ppb
- Trace Impurities - Zinc (Zn):≤200.0 ppb
Specification Test Results
About this item
:
:98.08 g/mol
UN:1830
ADR:8,II
:GHS05
:GHS07
:GHS08
CAS Number:7664-93-9
Formula:H₂SO₄
MDL Number:MFCD00064589