Order Entry
Austria
ContactUsLinkComponent
Sulphuric acid 50% (w/w), BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Corrosive
Irritant
Health
Sulphuric acid 50% (w/w), BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Catalog # 9696-09
Supplier:  Avantor
CAS Number:  
undefined
Corrosive
Irritant
Health
Sulphuric acid 50% (w/w), BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Catalog # 9696-09
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    PE drum, black
  • Pk:
    600 lb
  • Seq:
    7350

Specifications

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
    49 - 51 %
  • Assay (as H₂SO₄) (by acidimetry):
    49 - 51 %
  • Chloride (Cl):
    ≤1 ppm
  • Phosphate (PO₄):
    ≤1 ppm
  • Trace Impurities - Heavy Metals:
    ≤0.5 ppm
  • Trace Impurities - Iron (Fe):
    ≤0.5 ppm

Specification Test Results

About this item

 :
 :98.08 g/mol
Boiling Pt:∼100 °C (1013 hPa)
Density:1.1…1.4 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-639-5
UN:2796
ADR:8,II
 :GHS05
 :GHS07
 :GHS08
CAS Number:7664-93-9
Formula:H₂SO₄
MDL Number:MFCD00064589