


Sulphuric acid 96%, CMOS for microelectronic, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:7664-93-9
:
Formula:H₂SO₄
MDL Number:MFCD00064589
:98.08 g/mol
Merck Index:14,08974
Boiling Pt:∼335 °C (1013 hPa)
Melting Pt:3 °C
Density:1.84 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-639-5
UN:1830
ADR:8,II
:GHS05
:GHS08
:GHS07
Specification Test Results
- For Microelectronic Use:95.0 - 97.0 %
- Recommended Storage Conditions: 15° - 100°F:= 10
- Assay (H₂SO₄):95.0 - 97.0 %
- Color (APHA):≤10
- Residue after Ignition:≤2 ppm
- Chloride (Cl):≤0.1 ppm
- Nitrate (NO₃):≤0.2 ppm
- Phosphate (PO₄):≤0.3 ppm
- Trace Impurities - Aluminum (Al):≤50.0 ppb
- Arsenic and Antimony (as As):≤5.0 ppb
- Trace Impurities - Barium (Ba):≤10.0 ppb
- Trace Impurities - Beryllium (Be):≤10.0 ppb
- Trace Impurities - Bismuth (Bi):≤20.0 ppb
- Trace Impurities - Boron (B):≤10.0 ppb
- Trace Impurities - Cadmium (Cd):≤10.0 ppb
- Trace Impurities - Calcium (Ca):≤50.0 ppb
- Trace Impurities - Chromium (Cr):≤50 ppb
- Trace Impurities - Cobalt (Co):≤10.0 ppb
- Trace Impurities - Copper (Cu):≤10.0 ppb
- Trace Impurities - Gallium (Ga):≤10.0 ppb
- Trace Impurities - Germanium (Ge):≤10.0 ppb
- Trace Impurities - Gold (Au):≤20 ppb
- Heavy Metals (as Pb):≤200.0 ppb
- Trace Impurities - Iron (Fe):≤100.0 ppb
- Trace Impurities - Lead (Pb):≤20.0 ppb
- Trace Impurities - Lithium (Li):≤10.0 ppb
- Trace Impurities - Magnesium (Mg):≤50.0 ppb
- Trace Impurities - Manganese (Mn):≤10.0 ppb
- Trace Impurities - Mercury (Hg):≤5.0 ppb
- Trace Impurities - Molybdenum (Mo):≤10.0 ppb
- Trace Impurities - Nickel (Ni):≤10.0 ppb
- Trace Impurities - Niobium (Nb):≤10.0 ppb
- Trace Impurities - Potassium (K):≤50 ppb
- Trace Impurities - Silicon (Si):≤50 ppb
- Trace Impurities - Silver (Ag):≤10.0 ppb
- Trace Impurities - Sodium (Na):≤100.0 ppb
- Trace Impurities - Strontium (Sr):≤10.0 ppb
- Trace Impurities - Tantalum (Ta):≤10.0 ppb
- Trace Impurities - Thallium (Tl):≤20.0 ppb
- Trace Impurities - Tin (Sn):≤50 ppb
- Trace Impurities - Titanium (Ti):≤10.0 ppb
- Trace Impurities - Vanadium (V):≤10.0 ppb
- Trace Impurities - Zinc (Zn):≤50 ppb
- Trace Impurities - Zirconium (Zr):≤10.0 ppb
- Particle Count at point of fill - 0.5 µm and greater:≤150 par/ml
- Particle Count at point of fill - 1.0 µm and greater:≤25 par/ml
Specification Test Results
Related Information
Frequently Bought Together