Order Entry
Austria
ContactUsLinkComponent
D-(+)-Sucrose (beet-derived) NF, HPLE, multi-compendial parenteral grade low in endotoxins, J.T.Baker®
D-(+)-Sucrose (beet-derived) NF, HPLE, multi-compendial parenteral grade low in endotoxins, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
D-(+)-Sucrose (beet-derived) NF, HPLE, multi-compendial parenteral grade low in endotoxins, J.T.Baker®
Supplier:  Avantor
Certificates

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specification Test Results

  • Bulk Pharmaceutical Chemical:
    Passes Test
  • NF - Identification:
    Passes Test
  • NF - Appearance of Solution:
    Passes Test
  • NF - Color Value:
    ≤45
  • NF - Conductivity:
    ≤35 µS/cm
  • NF - Optical Rotation (+):
    66.3 - 67.0 Degree
  • NF - Reducing Sugars:
    Passes Test
  • NF - Sulfite (SO2):
    ≤10 ppm
  • NF - Loss on Drying at 105 ºC:
    ≤0.1 %
  • EP/BP - Identification A:
    Passes Test
  • EP/BP - Identification B:
    Passes Test
  • EP/BP - Identification C:
    Passes Test
  • EP/BP - Appearance of Solution:
    Passes Test
  • EP/BP - Conductivity, uS cm-1:
    ≤35
  • EP/BP - Optical Rotation (+):
    66.3 - 67.0 Degree
  • EP/BP - Colour Value:
    ≤45
  • EP/BP - Reducing Sugars:
    Passes Test
  • EP/BP - Sulfite (SO₂):
    ≤10 ppm
  • EP/BP - Loss on Drying at 105°C:
    ≤0.1 %
  • JP - Identification:
    Passes Test
  • JP - Optical Rotation (+):
    66.3 - 67.0 Degree
  • JP - Color Value:
    ≤45
  • JP - Clarity of Solution:
    Passes Test
  • JP - Dextrins:
    Passes Test
  • JP - Sulfite (SO₂):
    ≤10 ppm
  • JP - Conductivity, uS cm-1:
    ≤35
  • JP - Loss on Drying at 105°C:
    ≤0.1 %
  • JP - Reducing Sugars:
    Passes Test
  • Assay (HPLC):
    ≥99 %
  • Endotoxin Concentration (EU/g):
    ≤0.6
  • Microbial Testing - Salmonella (USP):
    None Detected
  • Microbial Testing - E. coli (USP):
    None Detected
  • Microbial Testing - Staphylococcus aureus (USP):
    None Detected
  • Microbial Testing - Pseudomonas aeruginosa (USP):
    None Detected
  • Microbial Testing - Total Yeast and Molds (USP):
    ≤100 cfu/g
  • Microbial Testing - Total Aerobic Microbial Count (USP):
    ≤100 cfu/g
  • Microbial Testing -Bile tolerant gram-negative bacteria:
    None Detected
  • Microbial Testing - Clostridia:
    None Detected
  • Microbial Testing - Candida albicans:
    None Detected
  • Trace Impurities - Aluminum (Al), For Information Only:
    For Information Only
  • Trace Impurities - Arsenic (As), For Information Only:
    For Information Only
  • Trace Impurities - Cadmium (Cd), For Information Only:
    For Information Only
  • Trace Impurities - Chromium (Cr), For Information Only:
    For Information Only
  • Trace Impurities - Copper (Cu), For Information Only:
    For Information Only
  • Trace Impurities - Iridium (Ir), For Information Only:
    For Information Only
  • Trace Impurities - Iron (Fe), For Information Only:
    For Information Only
  • Trace Impurities - Lead (Pb), For Information Only:
    For Information Only
  • Trace Impurities - Manganese (Mn), For Information Only:
    For Information Only
  • Trace Impurities - Mercury (Hg), For Information Only:
    For Information Only
  • Trace Impurities - Molybdenum (Mo), For Information Only:
    For Information Only
  • Trace Impurities - Nickel (Ni), For Information Only:
    For Information Only
  • Trace Impurities - Osmium (Os), For Information Only:
    For Information Only
  • Trace Impurities - Palladium (Pd), For Information Only:
    For Information Only
  • Trace Impurities - Platinum (Pt), For Information Only:
    For Information Only
  • Trace Impurities - Rhodium (Rh), For Information Only:
    For Information Only
  • Trace Impurities - Ruthenium (Ru), For Information Only:
    For Information Only
  • Trace Impurities - Vanadium (V), For Information Only:
    For Information Only
  • Trace Impurities - Zinc (Zn), For Information Only:
    = 3000 ppm
  • Residual Methanol:
    ≤3000 ppm
  • Residual Ethanol:
    ≤5000 ppm
  • Residual Isopropanol:
    ≤5000 ppm

Specification Test Results

Related Information

Frequently Bought Together