Order Entry
Austria
ContactUsLinkComponent
di-Sodium L(+)-tartrate dihydrate 99.0-101.0%, granular, AR® ACS, Macron Fine Chemicals™
di-Sodium L(+)-tartrate dihydrate 99.0-101.0%, granular, AR® ACS, Macron Fine Chemicals™
Catalog # 2386-20
CAS Number:  
undefined
di-Sodium L(+)-tartrate dihydrate 99.0-101.0%, granular, AR® ACS, Macron Fine Chemicals™
Catalog # 2386-20
CAS Number:  

Specifications

  • Pack type:
    Case, Mall STAKMOR
  • Pk:
    12 kg
  • Seq:
    4187

Specifications

Specification Test Results

  • Meets ACS Reagent Chemical Requirements:
  • For Laboratory,Research,or Manufacturing Use:
    99.0 - 101.0 %
  • ACS - Assay (Na₂C₄H₄O₆·2H₂O):
    99.0 - 101.0 %
  • ACS - Loss on Drying at 150°C:
    15.61 - 15.71 %
  • ACS - pH of 5% Solution at 25°C:
    7.0 - 9.0
  • ACS - Insoluble Matter:
    ≤0.005 %
  • ACS - Chloride (Cl):
    ≤5 ppm
  • ACS - Phosphate (PO₄):
    ≤5 ppm
  • ACS - Sulfate (SO₄):
    ≤0.005 %
  • ACS - Ammonium (NH₄):
    ≤0.003 %
  • ACS - Calcium (Ca):
    ≤0.01 %
  • ACS - Heavy Metals (as Pb):
    ≤5 ppm
  • ACS - Iron (Fe):
    ≤0.001 %

Specification Test Results

About this item

 :
 :230.08 g/mol
Merck Index:12,08833
Melting Pt:57 °C
Density:1.818 g/cm³ (20 °C)
EINECS:212-773-3
CAS Number:6106-24-7
Formula:NaOOC(CHOH)₂COONa·2H₂O
MDL Number:MFCD00150035