Sodium hydroxide solution 50%, Biotech Reagent, J.T.Baker®
: Avantor
: Caustic soda
:1310-73-2
:
:NaOH
:MFCD00003548
:40 g/mol
:1823
:8,II
- Environmentally Preferable:
- Assay (NaOH):49 - 51 %
- Appearance (clear, colorless liquid):passes test
- Potassium (K):max. 0.01 %
- Identification (Na):passes test
- Sodium Carbonate (Na2CO3):max. 1.0 %
- Trace Impurities (in ppm) Chlorides (as Cl):max. 5
- Trace Impurities (in ppm) Iron (Fe):max. 2
- Trace Impurities (in ppm) Heavy Metals (as Pb):max. 1
Frequently Bought Together