Order Entry
Austria
ContactUsLinkComponent
Sodium hydroxide 50% in aqueous solution, AR®, Macron Fine Chemicals™
Corrosive
Irritant
Sodium hydroxide 50% in aqueous solution, AR®, Macron Fine Chemicals™
Catalog # 7706-68
CAS Number:  
undefined
Corrosive
Irritant
Sodium hydroxide 50% in aqueous solution, AR®, Macron Fine Chemicals™
Catalog # 7706-68
CAS Number:  

Specifications

  • Pack type:
    IBC/tote
  • Pk:
    890 L
  • Seq:
    5729

Specifications

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
    49.5 - 52.0 %
  • Protect from air to avoid absorption of carbon dioxide.:
    Passes Test
  • Assay (NaOH):
    49.5 - 52.0 %
  • Identification:
    Passes Test
  • Ammonium Hydroxide Precipitate:
    ≤0.010 %
  • Chloride (Cl):
    ≤0.0025 %
  • Heavy Metals (as Ag):
    ≤0.0015 %
  • Iron (Fe):
    ≤0.0005 %
  • Nitrogen Compounds (as N):
    ≤0.0005 %
  • Potassium (K):
    ≤0.025 %
  • Phosphate (PO₄):
    ≤0.0005 %
  • Sulfate (SO₄):
    ≤0.0025 %
  • Sodium Carbonate (Na₂CO₃):
    ≤0.1 %

Specification Test Results

About this item

 :
 :40 g/mol
Density:1.53 g/cm³
UN:1824
ADR:8,II
 :GHS05
 :GHS07
CAS Number:1310-73-2
Formula:NaOH
MDL Number:MFCD00003548