Order Entry
Austria
ContactUsLinkComponent
Disodium hydrogen phosphate dihydrate, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Disodium hydrogen phosphate dihydrate, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Catalog # 0343.1000
Supplier:  Avantor
CAS Number:  
Disodium hydrogen phosphate dihydrate, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Catalog # 0343.1000
Supplier:  Avantor
CAS Number:  

Specifications

  • Pk:
    1 kg
  • Seq:
    4024

Specifications

Specification Test Results

  • Assay:
    min. 99 %
  • Chloride (Cl):
    max. 0.001 %
  • Iron (Fe):
    max. 0.001 %
  • Heavy Metals (as Pb):
    max. 0.001 %
  • Insoluble Matter:
    max. 0.005 %
  • pH of 0.1 M Solution at 25°C:
    9.0 - 9.5
  • Sulfate (SO4):
    max. 0.005 %

Specification Test Results

About this item

 :
 :177.99 g/mol
CAS Number:10028-24-7
Formula:Na₂HPO₄·2H₂O
MDL Number:MFCD00149182