Order Entry
Austria
ContactUsLinkComponent
Sodium azide ≥99.7%, OR, Macron Fine Chemicals™
Sodium azide ≥99.7%, OR, Macron Fine Chemicals™
Certificates
undefined
Sodium azide ≥99.7%, OR, Macron Fine Chemicals™
Certificates

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
    = 99.7 %
  • Assay (NaN₃):
    ≥99.7 %
  • Loss on Drying at 105°C:
    ≤0.1 %

Specification Test Results

Related Information

Frequently Bought Together