Order Entry
Austria
ContactUsLinkComponent
Sodium acetate trihydrate 99.0-101.0% (dried basis), crystals USP, Multi-Compendial, J.T.Baker®
Sodium acetate trihydrate 99.0-101.0% (dried basis), crystals USP, Multi-Compendial, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
Sodium acetate trihydrate 99.0-101.0% (dried basis), crystals USP, Multi-Compendial, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • Meets U.S.P Requirements:
    99.0 - 101.0%
  • GMP Manufactured Product:
    Passes Test
  • Meets F.C.C. Requirements:
    ≤0.035%
  • Food GMP Manufactured Product:
    ≤2.5
  • Meets J.P. Chemical Specifications:
    Passes Test
  • Meets E.P. Chemical Specifications:
    ≤0.05%
  • Meets B.P. Chemical Specifications:
    38.0 - 41.0%
  • Preserve in tight containers, store at room temperature.:
    7.5 - 9.2
  • CAUTION: For Manufacturing, processing or repackaging:
    Passes Test
  • Bulk Food Chemical:
    ≤0.005%
  • Bulk Pharmaceutical Chemical:
    99.0 - 101.0%
  • USP - Assay (CH₃COONa) (dried basis):
    99.0 - 101.0 %
  • USP - Calcium and Magnesium:
    Passes Test
  • USP - Chloride (Cl):
    ≤ 0.035 %
  • USP - Endotoxin Concentration (EU/g):
    ≤ 2.5
  • USP - Identification:
    Passes Test
  • USP - Insoluble Matter:
    ≤ 0.05 %
  • USP - Loss on Drying:
    38.0 - 41.0 %
  • USP - pH of 5% Solution at 25°C:
    7.5 - 9.2
  • USP - Potassium (K):
    Passes Test
  • USP - Sulfate (SO₄):
    ≤ 0.005 %
  • EP/BP - Assay (CH₃COONa):
    99.0 - 101.0 %
  • EP/BP - Identification A:
    Passes Test
  • EP/BP - Identification B:
    Passes Test
  • EP/BP - Identification C:
    Passes Test
  • EP/BP - Appearance of Solution:
    Passes Test
  • EP/BP - pH (1 in 20):
    7.5 - 9.0
  • EP/BP - Reducing Substances:
    Passes Test
  • EP/BP - Chloride (Cl):
    ≤ 200 ppm
  • EP/BP - Sulfate (SO₄):
    ≤ 200 ppm
  • EP/BP - Calcium and Magnesium:
    ≤ 50 ppm
  • EP/BP - Iron (Fe):
    ≤ 10 ppm
  • EP/BP - Loss on Drying:
    39.0 - 40.5 %
  • JP - Assay (CH₃COONa):
    99.5 - 101.0 %
  • JP - Identification:
    Passes Test
  • JP - Clarity and Color of Solution:
    Passes Test
  • JP - Acidity or Alkalinity:
    Passes Test
  • JP - Chloride (Cl):
    ≤ 0.011 %
  • JP - Sulfate (SO₄):
    ≤ 0.017 %
  • JP - Heavy Metals (as Pb):
    ≤ 10 ppm
  • JP - Calcium and Magnesium:
    Passes Test
  • JP - Arsenic (As):
    ≤ 2 ppm
  • JP - Substances Reducing Permanganate:
    Passes Test
  • JP - Loss on Drying:
    39.0 - 40.5 %
  • FCC - Alkalinity:
    ≤ 0.05 %
  • FCC - Assay (CH₃COONa) (dried basis):
    99.0 - 101.0 %
  • FCC - Identification A:
    Passes Test
  • FCC - Identification B:
    Passes Test
  • FCC - Lead (Pb):
    ≤ 2 mg/kg
  • FCC - Loss on Drying:
    38.0 - 41.0 %
  • FCC - Potassium Compounds:
    Passes Test

Specification Test Results

Related Information

Frequently Bought Together