Order Entry
Austria
ContactUsLinkComponent
Potassium dihydrogen phosphate 98.0-100.5% (dried basis) NF, FCC, J.T.Baker®
Irritant
Potassium dihydrogen phosphate 98.0-100.5% (dried basis) NF, FCC, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
Irritant
Potassium dihydrogen phosphate 98.0-100.5% (dried basis) NF, FCC, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • GMP Manufactured Product:
    98.0 - 100.5%
  • Meets N.F. Requirements:
    ≤3 ppm
  • Food GMP Manufactured Product:
    ≤10 mg/kg
  • Meets F.C.C. Requirements:
    Passes Test
  • CAUTION: For Manufacturing, processing or repackaging:
    Passes Test
  • Bulk Pharmaceutical Chemical:
    ≤0.2%
  • Bulk Food Chemical:
    ≤5 ppm
  • NF - Assay (KH₂PO₄) (dried basis):
    98.0 - 100.5 %
  • NF - Arsenic (As):
    ≤3 ppm
  • NF - Fluoride (F):
    ≤10 mg/kg
  • NF - Identification A:
    Passes Test
  • NF - Identification B:
    Passes Test
  • NF - Insoluble Substances:
    ≤0.2 %
  • NF - Lead (Pb):
    ≤5 ppm
  • NF - Loss on Drying at 105°C:
    ≤1.0 %
  • FCC - Assay (KH₂PO₄) (dried basis):
    98.0 - 100.5 %
  • FCC - Arsenic (As):
    ≤3 ppm
  • FCC - Fluoride (F):
    ≤10 mg/kg
  • FCC - Identification A:
    Passes Test
  • FCC - Identification B:
    Passes Test
  • FCC - Insoluble Substances:
    ≤0.2 %
  • FCC - Lead (Pb):
    ≤2 mg/kg
  • FCC - Loss on Drying at 105°C:
    ≤1.0 %

Specification Test Results

Related Information

Frequently Bought Together