Potassium nitrate 99.0-100.5%, granular USP, FCC, J.T.Baker®
3192-88
: Avantor
:
- Pack type:HDPE drum, Flowmor™ package
- Pk:26,25 kg
- Seq:4298
- Environmentally Preferable:
- Bulk Pharmaceutical Chemical:99.0 - 100.5 %
- USP - Assay (KNO₃):99.0 - 100.5 %
- USP - Arsenic (As):≤3 ppm
- USP - Chloride (Cl):≤0.03 %
- USP - Identification:Passes Test
- USP - Iron (Fe):≤0.001 %
- USP - Lead (Pb) (USP):≤0.001 %
- USP - Nitrite (NO₂):≤5 ppm
- USP - Sodium (Na):≤0.1 %
- USP - Sulfate (SO₄):≤0.1 %
- FCC - Assay (KNO₃) (dried basis):99.0 - 100.5 %
- FCC - Chlorate:Passes Test
- FCC - Identification:Passes Test
- FCC - Lead (Pb) (FCC):≤4 mg/kg
- FCC - Loss on Drying at 105°C:≤1 %
: Nitrate of Potash
:
:101.1 g/mol
:2.10 g/cm³
:1486
:5.1,III
:GHS03
:7757-79-1
:KNO₃
:MFCD00011409