Order Entry
Austria
ContactUsLinkComponent
Potassium nitrate ≥99.0%, crystals, BAKER ANALYZED® ACS, J.T.Baker®
Oxidizing
Potassium nitrate ≥99.0%, crystals, BAKER ANALYZED® ACS, J.T.Baker®
Catalog # 3190-80
Supplier:  Avantor
CAS Number:  
undefined
Oxidizing
Potassium nitrate ≥99.0%, crystals, BAKER ANALYZED® ACS, J.T.Baker®
Catalog # 3190-80
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    HDPE drum, Flowmor™ package
  • Pk:
    20 kg
  • Seq:
    4241

Specifications

Specification Test Results

  • Meets ACS Reagent Chemical Requirements:
  • For Laboratory,Research,or Manufacturing Use:
  • Assay (KNO₃):
    ≥99.0 %
  • Insoluble Matter:
    ≤0.005 %
  • pH of 5% Solution at 25°C:
    4.5 - 8.5
  • Chloride (Cl):
    ≤0.002 %
  • ACS- Nitrite (NO₂):
    ≤0.001 %
  • ACS - Sulfate (SO₄):
    ≤0.003 %
  • Calcium (Ca):
    ≤0.005 %
  • ACS - Magnesium (Mg):
    ≤0.002 %
  • Sodium (Na):
    ≤0.005 %
  • Iodate (IO₃):
    ≤5 ppm
  • Phosphate (PO₄):
    ≤3 ppm
  • Trace Impurities - ACS - Heavy Metals (as Pb):
    ≤5 ppm
  • Trace Impurities - Iron (Fe):
    ≤2.0 ppm

Specification Test Results

About this item

 :
 :101.1 g/mol
Merck Index:13,07733
Melting Pt:334 °C
Density:2.109 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-818-8
UN:1486
ADR:5.1,III
REACH:01-2119488224-35
 :GHS03
CAS Number:7757-79-1
Formula:KNO₃
MDL Number:MFCD00011409