Potassium iodide ≥99.0%, AR® ACS, free-flowing, Macron Fine Chemicals™
Catalog # 1123-20
Supplier: MACRON AVANTOR BRAND
CAS Number:
Specifications
- Pack type:Case, Mall STAKMOR
- Pk:12 kg
- Seq:4187
- Environmentally Preferable:
Specifications
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:≥99.0%
- ACS - Assay (KI):≥99.0 %
- ACS - Appearance (white, crystalline granules):Passes Test
- ACS - Barium (Ba):≤0.002 %
- ACS - Calcium (Ca):≤0.002 %
- ACS - Chloride and Bromide (as Cl):≤0.01 %
- ACS - Heavy Metals (as Pb):≤0.0005 %
- ACS - Insoluble Matter:≤0.005 %
- ACS - Iodate (IO₃):≤3 ppm
- ACS - Iron (Fe):≤0.0003 %
- ACS - Loss on Drying at 150°C:≤0.2 %
- ACS - Magnesium (Mg):≤0.001 %
- ACS - pH of 5% Solution at 25°C:6.0 - 9.2
- ACS - Phosphate (PO₄):≤0.001 %
- ACS - Sodium (Na):≤0.005 %
- ACS - Sulfate (SO₄):≤0.005 %
- Free Alkali, For Information Only:For Information Only
Specification Test Results
About this item
Synonyms: Dipotassium diiodide, Kali iodide
:
:166 g/mol
Merck Index:13,07727
Boiling Pt:1330 °C (1013 hPa)
Melting Pt:681 °C
Density:3.13 g/cm³ (20 °C)
:Management of Change (MOC) category = N
EINECS:231-659-4
:GHS07
:GHS08
Storage Temperature:Ambient
CAS Number:7681-11-0
Formula:Kl
MDL Number:MFCD00011405