Order Entry
Austria
ContactUsLinkComponent
Potassium hydroxide ≥85.0%, pellets, BAKER ANALYZED®, Electronic Grade, J.T.Baker®
Corrosive
Irritant
Potassium hydroxide ≥85.0%, pellets, BAKER ANALYZED®, Electronic Grade, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
Corrosive
Irritant
Potassium hydroxide ≥85.0%, pellets, BAKER ANALYZED®, Electronic Grade, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • For Microelectronic Use:
    ≥85.0%
  • Assay (KOH):
    ≥85.0 %
  • Potassium Carbonate (K₂CO₃):
    ≤1.0 %
  • Insoluble Matter:
    ≤50 ppm
  • Chloride (Cl):
    ≤0.01 %
  • Sulfate (SO₄):
    ≤20 ppm
  • Ammonium Hydroxide Precipitate:
    ≤0.02 %
  • Sodium (Na):
    ≤0.05 %
  • Nitrogen Compounds (as N):
    ≤10 ppm
  • Phosphate (PO₄):
    ≤5 ppm
  • Heavy Metals (as Ag):
    ≤10 ppm
  • Iron (Fe):
    ≤10 ppm
  • Mercury (Hg), For Information Only:
    ≤10 ppm
  • Nickel (Ni):
    ≤10 ppm
  • Calcium (Ca):
    ≤50 ppm

Specification Test Results

Related Information

Frequently Bought Together