Order Entry
Austria
ContactUsLinkComponent
 
Potassium chloride 99.0-100.5% (by argentometric titration), crystals, BAKER ANALYZED® ACS, J.T.Baker®
 :  Avantor
 
undefined
Potassium chloride 99.0-100.5% (by argentometric titration), crystals, BAKER ANALYZED® ACS, J.T.Baker®
 :  Avantor
 

 

  • Meets ACS Reagent Chemical Requirements:
    99.0 - 100.5%
  • For Laboratory,Research,or Manufacturing Use:
    ≤1.0%
  • Assay (KCl) (by Ag titrn):
    99.0 - 100.5 %
  • Loss on Drying at 105°C:
    ≤1.0 %
  • Insoluble Matter:
    ≤0.005 %
  • pH of 5% Solution at 25°C:
    5.4 - 8.6
  • Iodide (I):
    ≤0.002 %
  • Bromide (Br):
    ≤0.01 %
  • Calcium (Ca):
    ≤0.002 %
  • Chlorate and Nitrate (as NO₃):
    ≤0.003 %
  • Magnesium (Mg):
    ≤0.001 %
  • Nitrogen Compounds (as N):
    ≤0.001 %
  • Sulfate (SO₄):
    ≤0.001 %
  • Barium (Ba):
    Passes Test
  • Sodium (Na)(by FES):
    ≤0.005 %
  • Phosphate (PO₄):
    ≤5 ppm
  • Trace Impurities - ACS - Heavy Metals (as Pb):
    ≤5 ppm
  • Trace Impurities - Arsenic (As):
    ≤1 ppm
  • Trace Impurities - Iron (Fe):
    ≤2.0 ppm
  • Trace Impurities - Nickel (Ni):
    ≤1.000 ppm

 

Frequently Bought Together