Order Entry
Austria
ContactUsLinkComponent
 

Specifications

  • Pack type:
    Fibre drum
  • Pk:
    300 lb
  • Seq:
    7227
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Bulk Pharmaceutical Chemical:
    99.5 - 101.5 %
  • USP - Assay (KHCO₃) (dried basis):
    99.5 - 101.5 %
  • USP - Identification A:
    Passes Test
  • USP - Identification B:
    Passes Test
  • USP - Loss on Drying:
    ≤0.25 %
  • USP - Normal Carbonate:
    ≤2.5 %
  • FCC - Assay (KHCO₃) (dried basis):
    99.5 - 101.5 %
  • FCC - Identification A:
    Passes Test
  • FCC - Identification B:
    Passes Test
  • FCC - Carbonate:
    Passes Test
  • FCC - Lead (Pb):
    ≤2 mg/kg
  • FCC - Loss on Drying:
    ≤0.25 %
  • Arsenic (As):
    ≤3 ppm
  • Solubility:
    Passes Test
  • Appearance (white granules):
    Passes Test

Specification Test Results

About this item

Synonyms: Potassium bicarbonate
 :
 :100.12 g/mol
Merck Index:13,07691
Melting Pt:292 °C
Density:2.17 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:206-059-0
CAS Number:298-14-6
Formula:KHCO₃
MDL Number:MFCD00011402