Order Entry
Austria
ContactUsLinkComponent
Potassium hydrogen carbonate 99.7-100.5%, granular, AR® ACS, Macron Fine Chemicals™
Potassium hydrogen carbonate 99.7-100.5%, granular, AR® ACS, Macron Fine Chemicals™
Catalog # 6748-20
CAS Number:  
undefined
Potassium hydrogen carbonate 99.7-100.5%, granular, AR® ACS, Macron Fine Chemicals™
Catalog # 6748-20
CAS Number:  

Specifications

  • Pack type:
    Case, Mall STAKMOR
  • Pk:
    12 kg
  • Seq:
    4187
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Meets ACS Reagent Chemical Requirements:
  • For Laboratory,Research,or Manufacturing Use:
    99.7 - 100.5 %
  • ACS - Assay (KHCO₃):
    99.7 - 100.5 %
  • ACS - Ammonium (NH₄):
    ≤0.0005 %
  • ACS - Calcium (Ca):
    ≤0.002 %
  • ACS - Chloride (Cl):
    ≤0.001 %
  • ACS - Heavy Metals (as Pb):
    ≤0.0005 %
  • ACS - Insoluble Matter:
    ≤0.01 %
  • ACS - Iron (Fe):
    ≤5 ppm
  • ACS - Magnesium (Mg):
    ≤0.001 %
  • ACS - Phosphate (PO₄):
    ≤0.0005 %
  • ACS - Sodium (Na):
    ≤0.03 %
  • ACS - Sulfur Compounds (as SO₄):
    ≤0.003 %

Specification Test Results

About this item

Synonyms: Potassium bicarbonate
 :
 :100.12 g/mol
Merck Index:13,07691
Melting Pt:292 °C
Density:2.17 g/cm³ (20 °C)
 :Management of Change (MOC) category = N
Storage Temperature:Ambient
EINECS:206-059-0
CAS Number:298-14-6
Formula:KHCO₃
MDL Number:MFCD00011402