Potassium acetate 99.0-100.5% (dried basis), granular USP, Multi-Compendial endotoxin tested, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:127-08-2
:
Formula:CH₃COOK
MDL Number:MFCD00012458
:98.14 g/mol
Merck Index:13,07687
Melting Pt:304 °C
Density:1.57 g/cm³ (25 °C)
Flash Pt:>250 °C
Storage Temperature:Ambient
EINECS:204-822-2
Specification Test Results
- Meets E.P. Chemical Specifications:99.0 - 100.5%
- GMP Manufactured Product:Passes Test
- Meets B.P. Chemical Specifications:Passes Test
- Meets U.S.P Requirements:≤1.0%
- CAUTION: For Manufacturing, processing or repackaging:7.5 - 8.5
- Bulk Pharmaceutical Chemical:≤0.03%
- USP - Assay (CH₃COOK) (dried basis):99.0 - 100.5 %
- USP - Endotoxin Concentration (2.5 EU/g max.):Passes Test
- USP - Identification:Passes Test
- USP - Loss on Drying:≤1.0 %
- USP - pH (1 in 20):7.5 - 8.5
- USP - Residual Acetic Acid, For Information Only:7.5 - 9.0
- USP - Sodium (Na):≤0.03 %
- EP/BP - Assay (CH₃COOK) (dried basis):99.0 - 101.0 %
- EP/BP - Aluminum (Al):≤1 ppm
- EP/BP - Identification A:Passes Test
- EP/BP - Identification B:Passes Test
- EP/BP - Appearance of Solution:Passes Test
- EP/BP - pH (1 in 20):7.5 - 9.0
- EP/BP - Reducing Substances:Passes Test
- EP/BP - Chloride (Cl):≤200 ppm
- EP/BP - Sulfate (SO₄):≤200 ppm
- EP/BP - Iron (Fe):≤20 ppm
- EP/BP - Sodium (Na):≤0.50 %
- EP/BP - Loss on Drying:≤3.0 %
- Additional Analytical Control - Arsenic (As):≤8 ppm
Specification Test Results
Related Information
Frequently Bought Together