Order Entry
Austria
ContactUsLinkComponent
 
Potassium acetate, crystals, BAKER ANALYZED®, Electronic Grade, J.T.Baker®
Catalog # 6072-09
Supplier:  Avantor
CAS Number:  
undefined
Potassium acetate, crystals, BAKER ANALYZED®, Electronic Grade, J.T.Baker®
Catalog # 6072-09
Supplier:  Avantor
CAS Number:  

Specifications

  • Pk:
    175 lb
  • Seq:
    7194

Specifications

Specification Test Results

  • For Microelectronic Use:
    ≥ 99.0 %
  • Assay (CH₃COOK) (by non-aqueous titration):
    ≥99.0 %
  • Insoluble Matter:
    ≤0.005 %
  • pH of 5% Solution at 25°C:
    6.5 - 9.0
  • Chloride (Cl):
    ≤0.003 %
  • Phosphate (PO₄):
    ≤0.001 %
  • Sulfate (SO₄):
    ≤0.002 %
  • Calcium (Ca):
    ≤0.005 %
  • ACS - Magnesium (Mg):
    ≤0.002 %
  • Sodium (Na):
    ≤0.03 %
  • Trace Impurities - ACS - Heavy Metals (as Pb):
    ≤5 ppm
  • Trace Impurities - Iron (Fe):
    ≤5 ppm

Specification Test Results

About this item

 :
 :98.14 g/mol
Merck Index:13,07687
Melting Pt:304 °C
Density:1.57 g/cm³ (25 °C)
Flash Pt:>250 °C
Storage Temperature:Ambient
EINECS:204-822-2
CAS Number:127-08-2
Formula:CH₃COOK
MDL Number:MFCD00012458