Petroleum spirit, 35…60 °C, AR® ACS, Macron Fine Chemicals™
Catalog # 4980-26
Supplier: MACRON AVANTOR BRAND
CAS Number:
Specifications
- Pack type:Steel drum
- Pk:200 L
- Seq:5549
- Environmentally Preferable:
Specifications
Specification Test Results
- Meets ACS Reagent Chemical Requirements:Passes Test
- For Laboratory,Research,or Manufacturing Use:≤10
- ACS - Acidity:Passes Test
- ACS - Color (APHA):≤10
- ACS - Residue after Evaporation:≤0.001 %
- Appearance and Color:Passes Test
- ACS - Boiling Range(Lower):35 - 60 °C
- ACS - Boiling Range(Upper):35 - 60 °C
Specification Test Results
About this item
Synonyms: Gasoline, Petroleum ether, Petrol, Benzine
:
Boiling Pt:35…60 °C (1013 hPa)
EINECS:265-151-9
UN:1268
ADR:3,II
:GHS02
:GHS08
CAS Number:64742-49-0
:64742-82-1
MDL Number:MFCD00081849