Order Entry
Austria
ContactUsLinkComponent
Oxalic acid dihydrate 99.5-102.5%, crystals, AR® ACS, Macron Fine Chemicals™
Oxalic acid dihydrate 99.5-102.5%, crystals, AR® ACS, Macron Fine Chemicals™
Catalog # 2752-20
CAS Number:  
undefined
Oxalic acid dihydrate 99.5-102.5%, crystals, AR® ACS, Macron Fine Chemicals™
Catalog # 2752-20
CAS Number:  

Specifications

  • Pk:
    12 kg
  • Seq:
    4187
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Meets ACS Reagent Chemical Requirements:
  • For Laboratory,Research,or Manufacturing Use:
    99.5 - 102.5 %
  • ACS - Assay (H₂C₂O₄2H₂O):
    99.5 - 102.5 %
  • ACS - Calcium (Ca):
    ≤ 0.001 %
  • ACS - Chloride (Cl):
    ≤ 0.002 %
  • ACS - Heavy Metals (as Pb):
    ≤ 0.0005 %
  • ACS - Insoluble Matter:
    ≤ 0.005 %
  • ACS - Iron (Fe):
    ≤ 0.0002 %
  • ACS - Nitrogen Compounds (as N):
    ≤ 0.001 %
  • ACS - Residue after Ignition:
    ≤ 0.01 %
  • Solubility:
    Passes Test
  • ACS - Substances Darkened by Hot H₂SO₄:
    Passes Test
  • ACS - Sulfate (SO₄):
    ≤ 0.005 %

Specification Test Results

About this item

Synonyms: Ethanedioic acid dihydrate
 :
 :126.07 g/mol
UN:3261
ADR:8,III
CAS Number:6153-56-6
Formula:HO₂CCO₂H·2H₂O
MDL Number:MFCD00149102